Orders LinkContactUsLinkComponent
Polypropylene Buchner Funnels
Polypropylene Buchner Funnels
Polypropylene Buchner Funnels
Certificates
Polypropylene Buchner Funnels
Polypropylene Buchner Funnels
2 Options Available Below
Product Details & Documents
The polypropylene Buchner funnels are two-piece, light-weight, and easy-to-clean.
They are fully autoclavable. The top portion snaps to the bottom forming a vacuum tight seal.
Product Family Options
Información del producto
  • Top Diameter
    Availability
    Price
  • 100 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $220.35
    $220.35
    Log in Para disponibilidad
    Paquete de 10 (1UN)
    Paquete de 10 (1UN) - $240.19
    $240.19
    Each (1UN)
       $220.35
    Paquete de 10 (1UN)
       $240.19
    Supplier Number: BFP100
  • 125 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $293.45
    $293.45
    Log in Para disponibilidad
    Paquete de 6 (1UN)
    Paquete de 6 (1UN) - $319.86
    $319.86
    Each (1UN)
       $293.45
    Paquete de 6 (1UN)
       $319.86
    Supplier Number: BFP125