Orders LinkContactUsLinkComponent
Probes and Seekers
Probes and Seekers
Certificates
Probes and Seekers
Probes and Seekers
2 Options Available Below
Product Details & Documents
Easily separate structures.
  • Hexagonal handle
Used to track vessels, nerves, and muscle fibers during dissection. Probes and seekers are a core dissection instrument. The blunt end causes minimal damage to nearby structures, while the pointed end can be used to tease apart tissue for access.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Información del producto
  • Material
    Length
    Availability
    Price
  • 470315-196
    Clark Probe, Straight / Bent
    Clark Probe, Straight / Bent
    Stainless steel
    152 mm (6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $41.92
    $41.92
    Log in Para disponibilidad
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $45.69
    $45.69
    Each (1UN)
       $41.92
    Caja de 50 (1UN)
       $45.69
    Supplier Number: 35ANG
  • 470315-198
    Hubber Probe, Bent
    Hubber Probe, Bent
    Stainless steel
    152 mm (6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $45.69
    $45.69
    Log in Para disponibilidad
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $49.81
    $49.81
    Each (1UN)
       $45.69
    Caja de 50 (1UN)
       $49.81
    Supplier Number: 35H