Orders LinkContactUsLinkComponent
Iris Scissors
Iris Scissors
Iris Scissors
Certificates
Iris Scissors
2 Options Available Below
Product Details & Documents
For fine dissections.
  • Used for very fine dissections, and working in small areas
The blades are very narrow allowing for a very clean cut, and a short blade length gives greater control. These iris scissors have excellent rust resistance. The scissors marked TC, are tungsten carbon tipped for greater edge retention and strength.
Ordering information: Available in curved or straight configuration.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Length
    Material
    Curvature
    Availability
    Price
  • 470315-164
    Iris Scissors
    Iris Scissors
    115 mm (4.5")
    Stainless steel
    Curved
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $49.99
    $49.99
    Log in for availability
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $54.49
    $54.49
    Each (1UN)
       $49.99
    Caja de 25 (1UN)
       $54.49
    Supplier Number: 10
  • 470315-234
    TC Iris Scissors
    TC Iris Scissors
    115 mm (4.5")
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $927.11
    $927.11
    Log in for availability
    Caja de 20 (1UN)
    Caja de 20 (1UN) - $1,010.55
    $1,010.55
    Each (1UN)
       $927.11
    Caja de 20 (1UN)
       $1,010.55
    Supplier Number: DR9TC