Orders LinkContactUsLinkComponent
Fixed Blade Scalpels
Fixed Blade Scalpels
Fixed Blade Scalpels
Certificates
Fixed Blade Scalpels
Fixed Blade Scalpels
3 Options Available Below
Product Details & Documents
Secure blade attachment.
  • Stainless steel construction
  • Fixed blade for additional strength
Fixed blade scalpels typically are stronger and less likely break than disposable blades. Often used for heavy cutting jobs, fixed blades are also less likely to twist while cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Información del producto
  • Material
    Availability
    Price
  • 470315-172
    Microdissection Scalpel
    Microdissection Scalpel
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $66.66
    $66.66
    Log in Para disponibilidad
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $72.66
    $72.66
    Each (1UN)
       $66.66
    Caja de 25 (1UN)
       $72.66
    Supplier Number: 52
  • 470315-176
    Haris Scalpel
    Haris Scalpel
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $77.39
    $77.39
    Log in Para disponibilidad
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $84.36
    $84.36
    Each (1UN)
       $77.39
    Caja de 25 (1UN)
       $84.36
    Supplier Number: 56
  • 470315-248
    Narrow Blade Scalpel
    Narrow Blade Scalpel
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $53.21
    $53.21
    Log in Para disponibilidad
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $58.00
    $58.00
    Each (1UN)
       $53.21
    Caja de 50 (1UN)
       $58.00
    Supplier Number: 25