Orders LinkContactUsLinkComponent
Surgical Style Scissors
Surgical Style Scissors
Surgical Style Scissors
Certificates
Surgical Style Scissors
Surgical Style Scissors
6 Options Available Below
Product Details & Documents
Excellent for larger dissections.
  • Stainless steel construction
  • Variety of sizes and tip configurations
Of the many instruments used for dissection, scissors are the most common and versatile. These surgical style scissors will perform nearly all the functions you need for a dissection. The long handles and round finger loops, provide good leverage and excellent control using either hand. The Sharp/Sharp tip configuration works great for delicate or confined cutting while removing muscle groups, or blood vessels. The sharp/blunt configuration helps protect hidden tissues from accidental damage from a sharp tip, or when cutting materials like cartilage that require a heavier blade.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Información del producto
  • Length
    Material
    Blade Type
    Curvature
    Availability
    Price
  • 470315-216
    Scissors
    Scissors
    140 mm (5.5")
    Stainless steel
    Sharp/Blunt
    Curved
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $63.42
    $63.42
    Log in Para disponibilidad
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $69.12
    $69.12
    Each (1UN)
       $63.42
    Caja de 25 (1UN)
       $69.12
    Supplier Number: 5SBCD
  • 470315-160
    Scissors
    Scissors
    140 mm (5.5")
    Stainless steel
    Sharp/Sharp
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $59.12
    $59.12
    Log in Para disponibilidad
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $64.44
    $64.44
    Each (1UN)
       $59.12
    Caja de 25 (1UN)
       $64.44
    Supplier Number: 5SS
  • 470315-210
    Scissors
    Scissors
    115 mm (4.5")
    Stainless steel
    Sharp/Blunt
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $55.37
    $55.37
    Log in Para disponibilidad
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $60.36
    $60.36
    Each (1UN)
       $55.37
    Caja de 25 (1UN)
       $60.36
    Supplier Number: 4SB
  • 470315-214
    Scissors (Surgical)
    Scissors (Surgical)
    140 mm (5.5")
    Stainless steel
    Sharp/Blunt
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $60.71
    $60.71
    Log in Para disponibilidad
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $66.18
    $66.18
    Each (1UN)
       $60.71
    Caja de 25 (1UN)
       $66.18
    Supplier Number: 5SB
  • 470315-218
    Scissors
    Scissors
    165 mm (6.5")
    Stainless steel
    Sharp/Blunt
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $67.19
    $67.19
    Log in Para disponibilidad
    Caja de 20 (1UN)
    Caja de 20 (1UN) - $73.23
    $73.23
    Each (1UN)
       $67.19
    Caja de 20 (1UN)
       $73.23
    Supplier Number: 6SB
  • 470315-220
    Scissors
    Scissors
    152 mm (6")
    Stainless steel
    Sharp/Sharp
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $67.19
    $67.19
    Log in Para disponibilidad
    Caja de 20 (1UN)
    Caja de 20 (1UN) - $73.23
    $73.23
    Each (1UN)
       $67.19
    Caja de 20 (1UN)
       $73.23
    Supplier Number: 6SS