Orders LinkContactUsLinkComponent
Hemostatic Forceps
Hemostatic Forceps
Hemostatic Forceps
Hemostatic Forceps
Precise and secure holding.
  • Stainless steel construction
  • Available in curved or straight points
  • One handed operation
For dissection use, hemostats are usually used to hold tissue and pull it out of the way. The locking mechanism in the handle also makes them useful as a clamp to either close off small tubing, or hold items together.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Description
    Material
    Length
    Curvature
  • Hemostat
    Stainless steel
    127 mm (5")
    Curved
    Each (1UN)
    Each (1UN) - $59.12
    $59.12
    Caja de 20 (1UN)
    Caja de 20 (1UN) - $64.44
    $64.44
    Each (1UN)
       $59.12
    Caja de 20 (1UN)
       $64.44
     : 45CD
  • Kelly forceps
    Stainless steel
    140 mm (5.5")
    Curved
    Each (1UN)
    Each (1UN) - $63.42
    $63.42
    Caja de 20 (1UN)
    Caja de 20 (1UN) - $69.12
    $69.12
    Each (1UN)
       $63.42
    Caja de 20 (1UN)
       $69.12
     : 46CD
  • Hemostat
    Stainless steel
    203 mm (8")
    Straight
    Each (1UN)
    Each (1UN) - $164.46
    $164.46
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $179.26
    $179.26
    Each (1UN)
       $164.46
    Caja de 25 (1UN)
       $179.26
     : 86M
  • Hemostat
    Stainless steel
    245 mm (10")
    Straight
    Each (1UN)
    Each (1UN) - $199.95
    $199.95
    Caja de 20 (1UN)
    Caja de 20 (1UN) - $217.95
    $217.95
    Each (1UN)
       $199.95
    Caja de 20 (1UN)
       $217.95
     : 87M