Orders LinkContactUsLinkComponent
T-Pins
T-Pins
Generally used to hold tissue to the dissection pan, T-pins are also good for marking structures.
  • Two sizes available
  • Nickel plated
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Description
    Material
    Length
  • 1.5" T-Pins
    Nickel Plated
    38 mm (1.5")
    Paquete de 6
    Paquete de 6 - $45.69
    $45.69
    Caja de 50 (6UN)
    Caja de 50 (6UN) - $49.81
    $49.81
    Paquete de 6
       $45.69
    Caja de 50 (6UN)
       $49.81
     : 42-6/PK
  • 2" T Pins
    Nickel Plated
    50 mm (2")
    Paquete de 100
    Paquete de 100 - $163.35
    $163.35
    Caja de 10 (100UN)
    Caja de 10 (100UN) - $178.05
    $178.05
    Paquete de 100
       $163.35
    Caja de 10 (100UN)
       $178.05
     : 42-100/PK