Orders LinkContactUsLinkComponent
Microdissection Forceps
Microdissection Forceps
Microdissection Forceps
Certificates
Microdissection Forceps
Microdissection Forceps
3 Options Available Below
Product Details & Documents
For extremely fine work.
  • Highly precise forceps
  • High quality stainless steel construction
High quality materials and excellent craftsmanship combine in these microdissection forceps. Available as a straight fine point, a curved, or a self locking these forceps will exceed your expectations.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Material
    Length
    Curvature
    Availability
    Price
  • 470315-166
    Fine Point Forceps
    Fine Point Forceps
    Stainless steel
    115 mm (4.5")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $56.99
    $56.99
    Log in for availability
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $62.11
    $62.11
    Each (1UN)
       $56.99
    Caja de 50 (1UN)
       $62.11
    Supplier Number: 18
  • 470315-184
    Curved Forceps
    Curved Forceps
    Stainless steel
    115 mm (4.5")
    Curved
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $58.05
    $58.05
    Log in for availability
    Caja de 50 (1UN)
    Caja de 50 (1UN) - $63.28
    $63.28
    Each (1UN)
       $58.05
    Caja de 50 (1UN)
       $63.28
    Supplier Number: 18C
  • 470315-186
    Self Closing Forceps
    Self Closing Forceps
    Stainless steel
    115 mm (4.5")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $64.48
    $64.48
    Log in for availability
    Caja de 25 (1UN)
    Caja de 25 (1UN) - $70.29
    $70.29
    Each (1UN)
       $64.48
    Caja de 25 (1UN)
       $70.29
    Supplier Number: 18SL