Microdissection Forceps Set
Microdissection Forceps Set, Stainless steel
Catalog # 470315-242
Supplier: DR INSTRUMENTS INC. SE
Número de proveedor #: MDFKIT-1
Jump to:
Product Details & Documents
Precision cut points.
- Includes five varied tip 4.5 in forceps
- Extra fine points
- Storage case
Micro-dissection forceps set contains widely used five pairs of forceps. Each pair of forceps have highly precise and extra fine tips (points) to perform micro-dissection and or any fine work under the microscope. This kit includes two curved and three straight micro-dissection forceps
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Microdissection Forceps Set
- Material:Stainless steel