$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
Gram color staining kit
Gram color staining kit stabilized, BBL™ for bacteriology
Gram color staining kit stabilized, BBL™ for bacteriology
90000-708
:
BECTON DICKINSON DE MEXICO, S. MX
:
Gram color staining kit stabilized, BBL™ for bacteriology
90000-708
:
BECTON DICKINSON DE MEXICO, S. MX
:
212539
:
Tamaño:
250 ml (8.4 oz.)
Referencia:
90000-708
Nº de proveedor:
212539
: