Vertical Pipette Stand
Vertical Pipette Stand
Catalog # 470314-026
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0594
Jump to:
Product Details & Documents
The top disc that can be fixed on the rod has holes for holding pipettes while the lower one has pilot holes where the pipette tips rest and liquids drain down to the base. These discs can be adjusted on the rod according to the length of the pipette. The arrows provided on the discs should be kept parallel to ensure that corresponding holes are being centered.
Especificaciones
- Description:Vertical Pipette Stand