Squibb Dropping Funnels
Socket Size 24/29, 500 ml
Catalog # 470314-008
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH0476C
Jump to:
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Especificaciones
- Capacity:500 ml
- Description:Socket Size 24/29