Orders LinkContactUsLinkComponent
VWR® Gel-Loading Pipette Tips
Round MBP Style, Ø 0.57 mm, 200 µl, 68 mm, Bulk
  76321-562
VWR® Gel-Loading Pipette Tips
  76321-562
VWR® Gel-Loading Pipette Tips
VWR® Gel-Loading Pipette Tips
Round MBP Style, Ø 0.57 mm, 200 µl, 68 mm, Bulk
$1,419.35
Bolsa de 1,000 Case de 10 (1,000,UN)
VWR® Gel-Loading Pipette Tips faciliate precision sample loading into gels.
  • Standard 96 tip format is multi-channel friendly
  • Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available)
  • Sterile product is also pyrogen-free
Universal fit to accommodate most pipettes except Rainin LTS and Thermo Clip-Tip.
 : Choose from sterile or nonsterile formats, racked or bulk packaging, slim or MBP body styles.
  • Volume:
    200 µl
  • Length:
    68 mm
  • Packed:
    Bulk
  • Tip Style:
    Round MBP Style, Ø 0.57 mm