Ohaus® Compass Scales
About this item
Portable, stackable electronic scale with backlit LCD display, auto shutoff and stainless steel pan.
- Stackable storage
- LCD display with low battery indicator
- 3 AA batteries (included) or AC adapter (not included)
This portable electronic scale features up to 1000 hours of battery life, a large, backlit LCD display. Other features include energy-saving auto shut-off, low battery indicator, overload and underload indicators.
5 Options Available Below
Product Details & Documents
Portable, stackable electronic scale with backlit LCD display, auto shutoff and stainless steel pan.
- Stackable storage
- LCD display with low battery indicator
- 3 AA batteries (included) or AC adapter (not included)
This portable electronic scale features up to 1000 hours of battery life, a large, backlit LCD display. Other features include energy-saving auto shut-off, low battery indicator, overload and underload indicators.
Specifications for Product Family
Specifications
- Catalog No:
- 470313-236
- 470313-238
- 470313-240
- 470313-242
- 470313-244
- Model:
- CX221
- CX621
- CX1201
- CX2200
- CX5200
- Communication interface:
- RS232; USB
- RS232; USB
- RS232; USB
- RS232; USB
- RS232; USB
Specifications
Product Family Options
Product Information
- ModelWeighing CapacityPan/Platform SizeReadabilityAvailabilityPrice
- Supplier Number: 30428199Specifications:
More
Specifications
Cat. No.470313-236ModelCX221Communication interfaceRS232; USB - Supplier Number: 30428200Specifications:
More
Specifications
Cat. No.470313-238ModelCX621Communication interfaceRS232; USB - Supplier Number: 30428201Specifications:
More
Specifications
Cat. No.470313-240ModelCX1201Communication interfaceRS232; USB - Supplier Number: 30428202Specifications:
More
Specifications
Cat. No.470313-242ModelCX2200Communication interfaceRS232; USB - Supplier Number: 30428203Specifications:
More
Specifications
Cat. No.470313-244ModelCX5200Communication interfaceRS232; USB