Orders LinkContactUsLinkComponent
Glass Evaporating Basins
Glass Evaporating Basins
Glass Evaporating Basins
Certificates
Glass Evaporating Basins
Glass Evaporating Basins
7 Options Available Below
Product Details & Documents
Ideal for drying applications in microwave and ovens.
Evaporating basins made of borosilicate glass. The basins have a flat bottom with spout for easy pouring.
Eisco Labs borosilicate glass has the following heat tolerances:
Strain Point: –515 °C
Annealing Point: 565 °C
Softening Point: 820 °C
Product Family Options
Información del producto
  • Capacity
    Ø
    Height
    Availability
    Price
  • 15 ml
    0.9" (Base), 2" (Top)
    1.2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $228.40
    $228.40
    Each (1UN)
       $228.40
    Supplier Number: CH0068A01
  • 45 ml
    1.2" (Base), 2.4" (Top)
    1.9"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $283.77
    $283.77
    Each (1UN)
       $283.77
    Supplier Number: CH0068A02
  • 100 ml
    1.4" (Base), 3.1" (Top)
    1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $406.85
    $406.85
    Each (1UN)
       $406.85
    Supplier Number: CH0068A
  • 250 ml
    3"
    1.3"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,451.66
    $1,451.66
    Each (1UN)
       $1,451.66
    Supplier Number: CH0068B
  • 500 ml
    2.3" (Base), 5.2" (Top)
    2.5"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $735.23
    $735.23
    Each (1UN)
       $735.23
    Supplier Number: CH0068B01
  • 600 ml
    2.3" (Base), 5.9" (Top)
    3.4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.24
    $1,216.24
    Each (1UN)
       $1,216.24
    Supplier Number: CH0068C
  • 2000 ml
    3.6" (Base), 8" (Top)
    4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,837.00
    $1,837.00
    Each (1UN)
       $1,837.00
    Supplier Number: CH0068D