Polar Silica-Based SPE Columns, Honeywell Burdick & Jackson
Catálogo # BJ9056
Proveedor: HONEYWELL SPECIALTY CHEMICALS MX
Product available on GSA Advantage®
Especificaciones
- Tamaño de partícula:40 - 63 µm
- Tamaño de poro:60 Å
- Capacidad:7.0 ml
- Environmentally Preferable:
- Extremo tapado:N/A
- Surface area:500 m²/g
- Bed Weight:500 mg
- Referencia:BJ9056
- Packing Material:Silica
- Phase:Silica (Si)
- Nº de proveedor:9056
Especificaciones
Acerca de este artículo
These SPE columns are made from medical-grade polypropylene with PE frits.
- Wide selection of support materials and multiple cartridge/column sizes
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- Reliable quality
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar.