Aluminum / Stainless Steel Laboratory Jacks
Catalog # 470313-328
Supplier: GSC INTERNATIONAL SE
Specifications
- Descripción:150 mm Platform
- Referencia:470313-328
Specifications
About this item
These aluminum and stainless-steel laboratory jacks provide a helpful platform for use in the lab.
Two sizes to choose from:
150 mm (~5.9") by 150 mm Platform, Maximum Height of 260 mm (~10.25"), Holds up to 25 kg (~55lbs.)
200 mm (~7.9") by 200 mm Platform, Maximum Height of 280 mm (~11"), Holds up to 30 kg (~66lbs.)