12-Piece Cork Borer Set
Catalog # 470313-326
Supplier: GSC INTERNATIONAL SE
Specifications
- Ø orificio:4.8 - 21 mm
- Descripción:12-piece cork borer set
- Referencia:470313-326
Specifications
About this item
The cork borer set includes 12 borers, in sizes ranging from 4.8 to 21 mm.
- Borers are made of nickel-plated steel
- Ergonomic handles provide a good grip on the tool
- Hole diameters range between 4.8 and 21 mm