pH Buffer Solutions, Referenced at 20 °C, Traceable to NIST, Fluka™, Honeywell Research Chemicals
Catálogo # BJ31044-1L
Supplier: HONEYWELL INTERNATIONAL INC MX
CAS Number:
Specifications
- Tipo de env.:Plastic bottle
- Tamaño:1x 1 L
- pH Buffer Type:HCl/KCl
- Referencia:BJ31044-1L
- pH:1.0
- Certificación:Traceable to SRM from NIST
- pH, precisión:±0.02
Specifications
About this item
- Citrate and phosphate buffer solutions contain fungicide
- Traceable to SRM from NIST