Sample Spoons, Bürkle
Sample Spoon, 210 mm (8.27"), Stainless steel, 2 ml (0.07 oz.)
Catalog # 76319-782
Supplier: BURKLE INC
Supplier #: 5320-0210
Jump to:
Product Details & Documents
Sampling Spoons are designed for withdrawing small quantities of samples.
- Made of AISI 304 (1.4301) stainless steel
- Largely chemical resistant (not against acids and alkalis)
This specially shaped spoon allows users to reach almost every corner in a container, while preventing samples from trickling out.
Specifications
- Capacity:2 ml (0.07 oz.)
- Description:Sample Spoon
- Length:210 mm (8.27")
- Material:Stainless steel