Buckets, Stainless Steel, Bürkle
Catalog # 76319-764
Supplier: BURKLE INC
Specifications
- Capacidad:10 L (2.6 gal.)
- Descripción:Bucket, stainless steel
- Altura:275 mm (10.8")
- Referencia:76319-764
- Diámetro:275 mm (10.83")
Specifications
About this item
The bucket is made entirely of high-gloss polished stainless steel AISI 304 (1.4301).
- Smooth stainless steel surface makes it easy to clean
- Rounded and movable handle makes it comfortable to move the bucket
Suitable for moving liquids and bulk materials properly, especially in chemical, pharmaceutical, or cosmetic industries, as well as the food and animal feed industries.