VWR® Sterile T-Shaped Cell Spreaders
Catálogo # 76299-566
Proveedor: VWR International
Especificaciones
- Esterilidad:Sterile
- Color:Purple
- Descripción:T-Shape Spreader
- Referencia:76299-566
- Empaquetado:10/Peel Bag
Especificaciones
Acerca de este artículo
VWR® disposable T-shaped Cell Spreaders are ideal for spreading bacterial cultures evenly.
- Spread and disperse liquids across the surface of culture plates
- Smooth edges minimize 'gouging'
- Sterilized by EO irradiation
- BSE/TSE-free, and latex-free
VWR® T-Shaped Cell Spreaders are manufactured under EN ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certificaciones: ISO 13485 :2003 and ISO 9001: 2008.