Polyproplyene Test Tube Drying Racks
Test Tube Drying Rack, 50 Places for up to 16 mm Tubes, Pack of 6
Catalog # 470164-058
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: 77852
Jump to:
Product Details & Documents
Dry up to 50 tubes at once.
- Autoclavable
- 5×10 array of pegs
- Conveniently sold in package of 6 racks
These polypropylene racks can conveniently hold inverted tubes for drying and to minimize collection of airborne contaminants. Rack can hold up to 50×16 mm diameter tubes. Racks can also be used to hold and dry electrophoresis and chromatography plates.
Especificaciones
- Description:Test Tube Drying Rack, 50 Places for up to 16 mm Tubes, Pack of 6