Orders LinkContactUsLinkComponent
CHEMets® Visual Kit, Detergents (anionic surfactants, MBAS method), Chemetrics
CHEMets® Visual Kit, Detergents (anionic surfactants, MBAS method), Chemetrics
Detergents CHEMets Refill, Methylene blue, 0 - 3 ppm
Catalog # CXR-9400
Supplier:   CHEMetrics
Supplier #:  R-9400
CHEMets® Visual Kit, Detergents (anionic surfactants, MBAS method), Chemetrics
Catalog # CXR-9400
Supplier:  CHEMetrics
CAS Number:  
CHEMets® Visual Kit, Detergents (anionic surfactants, MBAS method), Chemetrics
CHEMets® Visual Kit, Detergents (anionic surfactants, MBAS method), Chemetrics
Detergents CHEMets Refill, Methylene blue, 0 - 3 ppm
CAS Number:  
Price
$3,133.09
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for testing detergents (anionic surfactants, MBAS method) in water.
Delivery information: Kit comes in a cardboard box and contains everything needed to perform 20 tests: refill, comparator, reaction tube with lid, tip breaking tool, and instructions.
Especificaciones
Especificaciones
  • Chemical method:
    Methylene blue
  • Description:
    Detergents CHEMets Refill
  • Range:
    0 - 3 ppm
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 