Orders LinkContactUsLinkComponent
Volumetric Pipettes, Class A
Volumetric Pipettes, Class A
Certificates
Volumetric Pipettes, Class A
Volumetric Pipettes, Class A
4 Options Available Below
Product Details & Documents
Reusable Class-A pipettes made from high quality borosilicate 3.3 glass.
  • Color coded for easy identification
  • Single graduation mark
  • Calibrated for delivery
  • Autoclavable
These Class-A volumetric pipettes are perfect for everyday laboratory use. They are designed for transferring liquid substances from one vessel to another with high accuracy.
 : Calibrated according to DIN ISO 648 standards.
Product Family Options
Información del producto
  • Capacity
    Color Code
    Tolerance
    Availability
    Price
  • 1 ml
    Blue
    ±0.0007 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $197.25
    $197.25
    Each (1UN)
       $197.25
    Supplier Number: CH0559A
  • 2 ml
    Orange
    ±0.010 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $197.25
    $197.25
    Each (1UN)
       $197.25
    Supplier Number: CH0559B
  • 10 ml
    Red
    ±0.020 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $267.66
    $267.66
    Each (1UN)
       $267.66
    Supplier Number: CH0559D
  • 25 ml
    Blue
    ±0.030 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $307.97
    $307.97
    Each (1UN)
       $307.97
    Supplier Number: CH0559G