Specimen Containers with Snap Cap, Simport Scientific
Catalog # 10066-766
Supplier: SIMPORT SCIENTIFIC INC.
Specifications
- Ext.Ø:111 mm
- Esterilidad:Non sterile
- Capacidad:500 ml (17 oz.)
- Descripción:Specimen Container with Snap Cap
- Altura:75 mm
- Referencia:10066-766
- Nº de proveedor:C580-4
Specifications
About this item
Disposable containers for specimen collection.
- Cap and container are made of polypropylene
- Disposable
- Tight fitting lids
These disposable containers made of polypropylene are ideal for collection and storage of fluids, powders, solids, pathology specimens, and hazardous samples.