ChemZoil NL-339 Heavyweight Chemical Protection Gloves, Mapa Professional
32916-090
- Color del guante:Black
- Tamaño del guante:10
- Ambidiestro/Específico para cada mano:Específico para cada mano
- Desechable / Reutilizable:Reutilizable
- Material:Neopreno
- Longitud:14"
- Grosor:0.05"
- Material del revestimiento:Knit cotton
- Acabado:Pebbled
- Referencia:32916-090
- Nº de proveedor:339420
These gloves are designed for long term wear while handling high-risk chemicals providing comfort and protection.
- Pebbled embossing, for better grip on wet and slippery objects
- Excellent mechanical resistance and longer service life
- Thermal insulation from the cotton-knit lining for handling hot liquids
- Designed using anatomical shape
It provides chemical resistance against acids and alkalis, such as concentrated hydrofluoric acid, oils and greases, due to its neoprene multi-layer construction.
The pre-curved fingers provide unrivalled freedom of movement, for this high-risk chemical protection glove.
: CE Cat. III 0334 marked. Head and Fire EN 407: X1XXXX. Chemical Protection EN 374: ABCJKL. Mechanical Hazard: EN 388: 3121.