Orders LinkContactUsLinkComponent
Cryo-Grip® Waterproof Gloves, Tempshield®
Cryo-Grip® Waterproof Gloves, Tempshield®
Supplier:   TEMPSHIELD INC
Cryo-Grip® Waterproof Gloves, Tempshield®
Supplier:  TEMPSHIELD INC
Certificates
Cryo-Grip® Waterproof Gloves, Tempshield®
2 Options Available Below
Product Details & Documents
Waterproof Cryo-Grip® gloves offer improved grip on small objects while protecting hands and arms when working in hazardous, ultra-cold environments.
  • Maximum thermal protection
  • Lightweight and comfortable
  • Excellent dexterity
  • Breathable and stay-dry lining
Gloves are designed to provide a high level of thermal protection where the exposure to cryogenic fluids might exist. Made from state-of-the-art materials, these waterproof cryogenic gloves are made to wick away moisture and keep wearer hands dry, to work in comfort for extended periods of time.
Where other gripping materials crack and break in ultra-low environments, the innovative Cryo-Grip® fabric will stay flexible, even in extreme conditions. The multi-layered construction allows for a maximum level of grip, dexterity, flexibility, and thermal protection - essential features when function is important and safety is critical.
Certifications: CE marked. Cat III. EN 420. EN 511. EN 388.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 76181-140
  • 76181-142
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
Especificaciones
Product Family Options
Información del producto
  • Glove size
    Glove Color
    Length
    Availability
    Price
  • 76181-140
    Mid-Arm Length
    Mid-Arm Length
    S
    Blue
    13.5 - 15.5"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,754.27
    $4,754.27
    Log in Para disponibilidad
    Case de 10 (1UN)
    Case de 10 (1UN) - $47,542.71
    $47,542.71
    Each (1UN)
       $4,754.27
    Case de 10 (1UN)
       $47,542.71
    Supplier Number: CGMASWP-CS
    Especificaciones:
    More
    Specifications
    Cat. No.
    76181-140
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 76181-142
    Mid-Arm Length
    Mid-Arm Length
    M
    Blue
    13.5 - 15.5"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,754.27
    $4,754.27
    Log in Para disponibilidad
    Case de 10 (1UN)
    Case de 10 (1UN) - $47,542.71
    $47,542.71
    Each (1UN)
       $4,754.27
    Case de 10 (1UN)
       $47,542.71
    Supplier Number: CGMAMWP-CS
    Especificaciones:
    More
    Specifications
    Cat. No.
    76181-142
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable