Orders LinkContactUsLinkComponent
LilyPad Sewable Electronics Kit
LilyPad Sewable Electronics Kit
LilyPad Sewable Electronics Kit
Catalog # 470311-306
Supplier:   SPARKLE LABS
Número de proveedor #:  SL032
LilyPad Sewable Electronics Kit
Catalog # 470311-306
Supplier:  SPARKLE LABS
LilyPad Sewable Electronics Kit
LilyPad Sewable Electronics Kit
LilyPad Sewable Electronics Kit
Price
$4,568.32
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
The LilyPad Sewable Electronics Kit lets you explore the wonderful world of electronic sewing (e-sewing) and e-textiles through a series of introductory projects using the LilyPad system.
Easy-to-follow diagrams and troubleshooting tips make this a great introductory resource for crafters and creatives.
You will learn how to sew basic circuits to light up LEDs, control them with buttons and switches and even experiment with a pre-programmed LilyMini circuit that reacts to ambient light levels. In addition to LilyPad LEDs and battery holders, the kit comes with two LilyPad ProtoSnap boards that let you explore the circuit before you sew the pieces into a project.
The full-color LilyPad Sewable Electronics Kit Guide (included) contains step-by-step instructions for using LilyPad pieces to create four complete sewable circuit projects with conductive thread.
Delivery information: Includes 1×LilyPad E-Sewing ProtoSnap; 1×LilyMini ProtoSnap; 2×LilyPad Rainbow LED Strip; 2x LilyPad Coin Cell Battery Holder - switched - 20 mm; 1×LilyPad Parts Box; 4×Felt Sheet - 9"×12"; 4×Felt Sheet - 9"×6"; 1×Pin Back; 1×Elastic Cord - 12"; 1×White Embroidery Thread; 1×White Sewing Thread; 1×Fiberfill Stuffing; 1×Needle Set; 2×Conductive Thread Bobbin - 30' (stainless steel); 2x Coin Cell Battery - 20 mm (CR2032); 4×Printed Project Templates; 1×LilyPad Sewable Electronics Kit Guidebook.
Especificaciones
  • Description:
    LilyPad Sewable Electronics Kit