Orders LinkContactUsLinkComponent
VWR® Urethane Lab Chairs
Supplier:   VWR International
VWR® Urethane Lab Chairs
Supplier:  VWR International
Certificates
VWR® Urethane Lab Chairs
2 Options Available Below
Product Details & Documents
These chairs have easy-to-clean seats and backrests which resist many chemicals, water, paint and more, combined with ergonomic functionality that make them a popular choice of laboratory professionals.
  • Other components include an easy-to-use ergonomic task control with seat height and back tilt adjustments and 26" Ø black composite base
  • Designed for laboratory applications, including those in healthcare, education and industrial settings
  • Backed by a 10-year warranty (lifetime on pneumatic)
Lab chairs Line features a refined yet pragmatic collection of chairs and stool designed for today laboratory professionals, applications and settings. The combination of european design with proven ergonomic functionality and engineering results in a focused set of models that meet real-world lab seating challenges.
 : Meets ANSI/BIFMA standards for durability and safety.
 : This product can expose you to chemicals including wood dust, which is known to the state of california to cause cancer. For more information go to www.p65warnings.ca.gov.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 76308-796
  • 76308-800
  • Model:
  • 76308-796
  • 76308-800
  • For use with:
  • Medium height benches (32.75 - 42")
  • Low benches (27-32")
  • Seat/back color:
  • Black
  • Black
  • Seat/back material:
  • Urethane
  • Urethane
  • Seat dimensions:
  • W18.5×D17×T1.5"
  • W18.5×D17×T1.5"
  • Backrest dimensions:
  • W×H 16.5×12.5"
  • W×H 16.5×12.5"
Especificaciones
Product Family Options
Información del producto
  • Color
    Foot Style
    Seat Height
    Availability
    Price
  • 76308-796
    Lab chair, for bench height
    Lab chair, for bench height
    Black
    20" dia. adjustable chrome footring/black spoke; dual soft-wheel casters
    20.75 - 30"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $19,044.85
    $19,044.85
    Each (1UN)
       $19,044.85
    Especificaciones:
    More
    Specifications
    Cat. No.
    76308-796
    Model
    76308-796
    For use with
    Medium height benches (32.75 - 42")
    Seat/back color
    Black
    Seat/back material
    Urethane
    Seat dimensions
    W18.5×D17×T1.5"
    Backrest dimensions
    W×H 16.5×12.5"
  • 76308-800
    Lab chair, for desk height
    Lab chair, for desk height
    Black
    No footring; dual soft-wheel casters
    15 - 20"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $15,525.24
    $15,525.24
    Each (1UN)
       $15,525.24
    Especificaciones:
    More
    Specifications
    Cat. No.
    76308-800
    Model
    76308-800
    For use with
    Low benches (27-32")
    Seat/back color
    Black
    Seat/back material
    Urethane
    Seat dimensions
    W18.5×D17×T1.5"
    Backrest dimensions
    W×H 16.5×12.5"