LaMotte® Soil Test Measuring Spoon
Soil Test Measuring Spoon
Catalog # 470225-860
Supplier: LA MOTTE CHEMICAL PROD CO SE
Número de proveedor #: 0699
Jump to:
Product Details & Documents
Soil test measuring spoon ideal for nitrate nitrogen test kit.
- Made of plastic
- 0.1 g capacity
Especificaciones
- Description:Soil Test Measuring Spoon