Acerca de este artículo
Ratiometric design in innovative, waterproof, handheld meters.
- Ratio measurement
- Waterproof to IP67
- Lithium ion rechargeable battery
- USB port
- 7 languages
- Backlit display
- Tungsten and Infrared
The multi-detector optical configuration assures long-term stability and minimizes stray light and color interference. The new ratiometric design allows for easy and accurate testing. The nephelometric mode measures 0-40 NTU/FNU, ratiometric mode 40-1000 NTRU/FNRU and 1000-2000 AU. Now pinpoint the range of interest with better, more reliable results. Ideally suited for low-level drinking water applications, mid-range industrial applications, and high-range environmental applications.
2020t version meets US EPA design criteria as specified by EPA 180.1, Rev. 2.0 (1993) and Standard Methods 2130 B-2001.
2020i version meets design criteria for quantitative methods of turbidity using optical turbidimeters as specified by ISO 7027.
Especificaciones
- Catalog No:
- 470308-442
- 470308-444
- Modelo:
- 2020t
- 2020i
- Fuente de luz:
- Tungsten lamp 2300 °K ±50 °K
- IR LED 860 nm ±10 nm, spectral bandwidth with 50 nm
- Detector:
- Photodiode, centered at 90° and 180°, maximum peak 400-600 nm
- Photodiode, centered at 90° and 180°
- Selección automática de rango:
- Yes
- Yes
- Rango de medición:
- 0-1,000 NTRU
0-17,500 ASBC
0-250 EBC - 0-1,000 FNRU
0-17,500 ASBC
0-250 EBC
- Memoria:
- Data logging 500 points
- Data logging 500 points
- Reproducibilidad:
- 0.02 NTRU or 1%
- 0.02 FNRU or 1%
- Resolución:
- 0–10.99 NTRU: 0.01
11.0–109.9 NTRU: 0.1
110–1000 NTRU: 1 - 0–10.99 FNRU: 0.01
11.0–109.9 FNRU: 0.1
110–1000 FNRU: 1
- W×D×H:
- 19.05×8.84×6.35 cm
- 19.05×8.84×6.35 cm
- Peso:
- 13 ounces
- 13 ounces