Laboratory Aprons
Supplier: UNITED SCIENTIFIC SUPPLIES
About this item
Protective laboratory apron for multipurpose use.
- Flexible
- Chemical resistant
- Apron includes cloth ties at the neck and waist
These aprons are made of a PVC-coated fabric which stays flexible. There is reinforcement at the points of strain.
4 Options Available Below
Product Details & Documents
Protective laboratory apron for multipurpose use.
- Flexible
- Chemical resistant
- Apron includes cloth ties at the neck and waist
These aprons are made of a PVC-coated fabric which stays flexible. There is reinforcement at the points of strain.
Specifications for Product Family
Specifications
- Catalog No:
- 470311-690
- 470311-692
- 470311-694
- 470311-696
- Garment application:
- Chemical protection|General Purpose|Laboratory
- Chemical protection|General Purpose|Laboratory
- Chemical protection|General Purpose|Laboratory
- Chemical protection|General Purpose|Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- PVC-coated fabric
- PVC-coated fabric
- PVC-coated fabric
- PVC-coated fabric
Specifications
Product Family Options
Product Information
- Garment fitL×WAvailabilityPrice
- Supplier Number: LBAP01Specifications:
More
Specifications
Cat. No.470311-690Garment applicationChemical protection|General Purpose|LaboratoryGenderUnisexFabricPVC-coated fabric - Supplier Number: LBAP02Specifications:
More
Specifications
Cat. No.470311-692Garment applicationChemical protection|General Purpose|LaboratoryGenderUnisexFabricPVC-coated fabric - Supplier Number: LBAP03Specifications:
More
Specifications
Cat. No.470311-694Garment applicationChemical protection|General Purpose|LaboratoryGenderUnisexFabricPVC-coated fabric - Supplier Number: LBAP04Specifications:
More
Specifications
Cat. No.470311-696Garment applicationChemical protection|General Purpose|LaboratoryGenderUnisexFabricPVC-coated fabric