Orders LinkContactUsLinkComponent
Ward's Base and Stand Sets
Ward's Base and Stand Sets
Ward's Base and Stand Sets
Certificates
Ward's Base and Stand Sets
Ward's Base and Stand Sets
1 Options Available Below
Product Details & Documents
These laboratory stands are the prefect addition to every general science laboratory and classroom.
  • Chemically resistant coating
  • Dual rod position
  • Versatilie laboratory stands
Perfect for setting up a physics experiment or titrating in the chemistry lab.
Product Family Options
Información del producto
  • Base Dimensions
    Rod Diameter
    Rod Length
    Availability
    Price
  • 470310-702
    Ward's Base and Stand Set
    Ward's Base and Stand Set
    100×150 mm
    11 mm
    50 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $603.03
    $603.03
    Each (1UN)
       $603.03
    Supplier Number: CH0656BSETA
  • 470310-704
    Ward's Premium Base and Stand
    Ward's Premium Base and Stand
    230×140 mm
    11 mm
    60 cm
    Discontinued
    Supplier Number: CH0656CSET