Single Scale Graduated Cylinders
Supplier: EISCO SCIENTIFIC LLC
Jump to:
Acerca de este artículo
Single Scale Graduated Cylinders with plastic base and protection collar.
- Class B
- Borosilicate glass gradauted cylinder
- Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
5 Options Available Below
Product Details & Documents
Single Scale Graduated Cylinders with plastic base and protection collar.
- Class B
- Borosilicate glass gradauted cylinder
- Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
Product Family Options
Información del producto
- CapacityGraduation IntervalAvailabilityPrice
- Supplier Number: CH0346A
- Supplier Number: CH0346B
- Supplier Number: CH0346D
- Supplier Number: CH0346C
- Supplier Number: CH0346E