Orders LinkContactUsLinkComponent
TPX Beakers
TPX Beakers
TPX Beakers
Certificates
TPX Beakers
8 Options Available Below
Product Details & Documents
These TPX (PMP) beakers are idea for general laboraotry use.
  • Chemically resistant
  • Autoclavable
  • Long lasting and high clarity beakers
Able to withstand temperatures up to 180 °C.
Product Family Options
Información del producto
  • Capacity
    Graduation Interval
    Graduation Range
    Availability
    Price
  • 50 ml
    5 ml
    5 - 50 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $255.28
    $255.28
    Each (1UN)
       $255.28
    Supplier Number: CH0138APR
  • 100 ml (3.4 oz.)
    10 ml
    10 - 100 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $319.26
    $319.26
    Each (1UN)
       $319.26
    Supplier Number: CH0138BPR
  • 250 ml
    25 ml
    25 - 250 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $511.65
    $511.65
    Each (1UN)
       $511.65
    Supplier Number: CH0138CPR
  • 600 ml
    50 ml
    100 - 600 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $614.32
    $614.32
    Each (1UN)
       $614.32
    Supplier Number: CH0138DPR
  • 1000 ml
    50 ml
    100 - 1000 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,062.01
    $1,062.01
    Each (1UN)
       $1,062.01
    Supplier Number: CH0138EPR
  • 2000 ml
    100 ml
    200 - 2000 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,726.28
    $1,726.28
    Each (1UN)
       $1,726.28
    Supplier Number: CH0138FPR
  • 5000 ml
    500 ml
    500 - 5000 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,795.47
    $3,795.47
    Each (1UN)
       $3,795.47
    Supplier Number: CH0138GPR
  • 50, 100, 250, 600, 1000 ml (Set)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,941.81
    $1,941.81
    Each (1UN)
       $1,941.81
    Supplier Number: CH0138PRSET