Glass Powder Funnels
Supplier: EISCO SCIENTIFIC LLC
Certificates
About this item
These funnels are available in a variety of different diameter openings.
- Borosilicate glass
- Plain 60° angle
- Fast and efficient
When pouring large particles into reaction vessels and beakers, powder funnels allow a clean and quick procedure. Powder bridging is minimized by a parallel, short stem.
Frequently Bought Together