Neoprene Stoppers
Supplier: EISCO SCIENTIFIC LLC
Jump to:
Acerca de este artículo
Versatile neoprene stoppers perfect for every laboratory.
- Solid, one hole or two hole stoppers are available
- Manufacturered according to sizes recommended by ASTM standards
Manufactured from long lasting neoprene rubbers. Suitable for use with petroleum products, mineral oils, fats as well as most inorganic acids and bases. Neoprene rubber is oil resistant and suitable for temperatures up to 120 °C (250 °F).
: Packs of 10.
46 Options Available Below
Product Details & Documents
Versatile neoprene stoppers perfect for every laboratory.
- Solid, one hole or two hole stoppers are available
- Manufacturered according to sizes recommended by ASTM standards
Manufactured from long lasting neoprene rubbers. Suitable for use with petroleum products, mineral oils, fats as well as most inorganic acids and bases. Neoprene rubber is oil resistant and suitable for temperatures up to 120 °C (250 °F).
: Packs of 10.
Product Family Options
Solid
Información del producto
- SizeAvailabilityPrice
- Supplier Number: CH0321A
- Supplier Number: CH0321B
- Supplier Number: CH0321C
- Supplier Number: CH0321D
- Supplier Number: CH0321E
- Supplier Number: CH0321F
- Supplier Number: CH0321G
- Supplier Number: CH0321H
- Supplier Number: CH0321I
- Supplier Number: CH0321J
- Supplier Number: CH0321K
- Supplier Number: CH0321L
- Supplier Number: CH0321N
- Supplier Number: CH0321O
- Supplier Number: CH0321P
- Supplier Number: CH0321Q
- Supplier Number: CH0321S
- Supplier Number: CH0321U
- Supplier Number: CH0321X
One Hole
Información del producto
- SizeAvailabilityPrice
- Supplier Number: CH0321A1H
- Supplier Number: CH0321D1H
- Supplier Number: CH0321E1H
- Supplier Number: CH0321F1H
- Supplier Number: CH0321G1H
- Supplier Number: CH0321H1H
- Supplier Number: CH0321J1H
- Supplier Number: CH0321K1H
- Supplier Number: CH0321L1H
- Supplier Number: CH0321M1H
- Supplier Number: CH0321N1H
- Supplier Number: CH0321P1H
- Supplier Number: CH0321Q1H
- Supplier Number: CH0321S1H
- Supplier Number: CH0321U1H
Two Holes
Información del producto
- SizeAvailabilityPrice
- Supplier Number: CH0321C2H
- Supplier Number: CH0321D2H
- Supplier Number: CH0321E2H
- Supplier Number: CH0321F2H
- Supplier Number: CH0321G2H
- Supplier Number: CH0321H2H
- Supplier Number: CH0321J2H
- Supplier Number: CH0321L2H
- Supplier Number: CH0321M2H
- Supplier Number: CH0321P2H
- Supplier Number: CH0321Q2H
- Supplier Number: CH0321U2H