Orders LinkContactUsLinkComponent
VWR® T Balances
VWR® T Balances
VWR® T Balances
VWR® T Balances
The VWR® T-Series balances feature state-of-the-art technology to provide the highest accuracy measurements.
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Internal and external calibration
  • Seven databases
  • 2× RS232, USB (A and B), ethernet and Wi-Fi communication interfaces
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
These balances provide accuracy and precision for any laboratory environment. Featuring internal calibration, IR proximity sensors for hands free operation, a multitude of communication options, and built in data protections, and provides advanced features for any application. The large, color touch screen allows for easy viewing of results and navigation through the balances menus. Data safety is provided via the internal memory allows for recording up to 500000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its 5-year warranty, this balance will provide reliable results for years to come.
 : C-UL (US) and CE.
 : Accessories not included (must be ordered separately).
 : The 0.01 and 0.1 mg readability models feature a draft shield (shield can't be removed). The 0.001 g readability feature a removable, glass shield. All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty.
  • Catalog No:
  • 76446-794
  • 76446-798
  • 76446-800
  • 76300-074
  • Model:
  • VWR-105TC
  • VWR-160TC
  • VWR-220TC
  • VWR-1003TC
  • Operating temperature range:
  • 10…40 °C (50…104 °F)
  • 10…40 °C (50…104 °F)
  • 10…40 °C (50…104 °F)
  • 10…40 °C (50…104 °F)
  • Plug type:
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Communication interface:
  • Wireless, 2×RS232, USB (A and B), Ethernet
  • Wireless, 2×RS232, USB (A and B), Ethernet
  • Wireless, 2×RS232, USB (A and B), Ethernet
  • Wireless, 2×RS232, USB (A and B), Ethernet
  • Voltage:
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
Product Family Options
3-Door draft shield
  • Model
    Weighing Capacity
    Linearity
    Pan/Platform Size
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
  • VWR-105TC
    120 g
    0.07 mg
    Ø 90 mm
    0.01 mg
    2.0 s
    0.02 mg (5% load)
    205.5W×351D×348W mm (8.1×13.8×13.7")
    Each (1UN)
    Each (1UN) - $192,698.16
    $192,698.16
    Each (1UN)
       $192,698.16
    Specifications
    Cat. No.
    76446-794
    Model
    VWR-105TC
    Operating temperature range
    10…40 °C (50…104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    Wireless, 2×RS232, USB (A and B), Ethernet
    Voltage
    100 - 240 V
  • VWR-160TC
    160 g
    0.2 mg
    Ø 100 mm
    0.1 mg
    2.0 s
    0.09 mg (5% load)
    205.5W×351D×348W mm (8.1×13.8×13.7")
    Specifications
    Cat. No.
    76446-798
    Model
    VWR-160TC
    Operating temperature range
    10…40 °C (50…104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    Wireless, 2×RS232, USB (A and B), Ethernet
    Voltage
    100 - 240 V
  • VWR-220TC
    220 g
    0.2 mg
    Ø 100 mm
    0.1 mg
    2.0 s
    0.09 mg (5% load)
    205.5W×351D×348W mm (8.1×13.8×13.7")
    Specifications
    Cat. No.
    76446-800
    Model
    VWR-220TC
    Operating temperature range
    10…40 °C (50…104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    Wireless, 2×RS232, USB (A and B), Ethernet
    Voltage
    100 - 240 V
Accessories
  •  
    Description
  • 10799-132
    VWR® Dot Matrix Printer
    Each (1UN) - $18,834.64
  •  
    Description
  • 10799-162
    Granite platform for weighing; 70 lbs. maximum load; 38.1W×33L cm (15×13") slab
    Each (1UN) - $42,199.68