Premiere® Positively Charged Slides
Catalog # 470303-814
Proveedor: C & A SCIENTIFIC CO INC
Especificaciones
- Descripción:Charged slides
- l×a:75×25 mm
- Grosor:1.0 mm
- Referencia:470303-814
Especificaciones
Acerca de este artículo
This slide is coated with a special adhesive formula (also known as "Positively Charged"), which allows the specimen to adhere to the slide easily, saving time and money in laboratory procedures.
- White-ended for clear labeling
- Pre-cleaned
- Corrosion-resistant glass