Premiere® Premium Grade Glass Microscope Slides
Catalog # 470303-812
Supplier: C & A SCIENTIFIC CO INC
Specifications
- Descripción:White-end slides
- l×a:75×25 mm
- Grosor:1.0 mm
- Referencia:470303-812
Specifications
About this item
Premiere® premium grade slides provide consistent results for all laboratory tests.
- Pre-cleaned
- Smoothly polished edges
- Corrosion-resistant glass
- Available plain, frosted-end or white-end formats
Premiere® brand slides are made of corrosion resistant glass with smoothly polished edges. These pre-cleaned slides measure 75×25mm, and are approximately 1 mm thick.