Orders LinkContactUsLinkComponent
Ward's Pipette Racks
Ward's Pipette Racks
Ward's Pipette Racks
Certificates
Ward's Pipette Racks
Ward's Pipette Racks
2 Options Available Below
Product Details & Documents
Protects and organizes glass and plastic pipettes.
  • Cut out viewing window
  • Available in magnetized option
Angled four shelved compartments keep 0.1 to 50 ml pipettes or cans within easy reach while preventing damage and clutter. Each pipette rack comes with two removeable attachments that hold pipette bulbs, fillers, and pumps. The Magnetized version has strong magnets on both sizes and mounts securely to most metal surfaces.
Product Family Options
Product Information
  • Dimensions
    Color
    Availability
    Price
  • 470311-172
    Ward's ABS pipette rack
    Ward's ABS pipette rack
    291×88×401 mm
    Clear
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,159.13
    $3,159.13
    Each (1UN)
       $3,159.13
    Supplier Number: HS20611W
  • 470311-174
    Ward's ABS magnetic pipette rack
    Ward's ABS magnetic pipette rack
    291×88×401 mm
    Blue
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,628.48
    $1,628.48
    Log in for availability
    Case de 10 (1UN)
    Case de 10 (1UN) - $1,775.05
    $1,775.05
    Each (1UN)
       $1,628.48
    Case de 10 (1UN)
       $1,775.05
    Supplier Number: HS20612MW