Cardboard Tube Storage Boxes and Partitions
A Low-cost way to organize tubes.
- Write-on surface for descriptions or self numbering systems
- Partitions come in two sizes
Economy cardboard racks are a quick and inexpensive solution with the added convenience of disposal after use.
Temperature : −191 °C
: Partitions are ordered separately and come in two convenient sizes.
: Partitions are supplied flat, packed for space-saving storage.
Frequently Bought Together