Ward's® Mini Magnifier
Mini magnifier, 16.1×13.5×20.7 cm
Catalog # 470311-198
Supplier: HEATHROW SCIENTIFIC SE
Número de proveedor #: HS15092D
Jump to:
Product Details & Documents
Mini magnifier enlarges objects for easier view.
- Swiveling, molded lens provides 1.75× magnification
- Excellent for viewing and counting colonies on bacterial plates
The Ward's® mini magnifier is mounted on a stand to keep hands free for manipulating samples. Excellent for viewing bacterial colonies on media, the magnifier pairs well with the Ward's® mini light box and Ward's® benchtop counter lines (both available separately).
Especificaciones
- Magnification:1.75×
- Description:Mini magnifier, 16.1×13.5×20.7 cm