LaMotte® TesTab® Nitrate Test Kit
Replacement glass test tube, with cap, graduated at 5 and 10 ml
Catalog # 470308-454
Supplier: LA MOTTE CHEMICAL PROD CO SE
Supplier #: 0898
LaMotte® TesTab® Nitrate Test Kit
Catalog # 470308-454
Supplier: LA MOTTE CHEMICAL PROD CO SE
CAS Number:
Jump to:
Product Details & Documents
Refill for The Comprehensive Water Monitoring Kit 470004-240 and LaMotte® TesTab® Water Investigation Kit 470007-270
- 100 Tests
- Range 0, 5, 20, 40 ppm (NH₃)
Each kit contains easy-to-read diagrammed instructions, a color chart, a test factor flash card and all the necessary apparatus and non-hazardous TesTabs® to perform 100 tests
Especificaciones
Especificaciones
- Description:Replacement glass test tube, with cap, graduated at 5 and 10 ml