3M™ Vinyl Foam Tapes
Supplier: 3M ELECTRONIC SPECIALTY MARKET
Certificates
About this item
These tapes are designed for demanding gasketing, cushioning, damping and insulating applications.
- Durable acrylic adhesive on one side of foam offers good aging and UV resistance
- Conformable properties allow taping or sealing on curved or irregular surfaces
- Designed to avoid stretching while leaving the liner in place
- Offers good aging characteristics
These tapes include a closed cell, high-density, flexible foam backing with adhesive which allows for quick-stick to a variety of substrates including many metals, glass and plastics. The silicone treated white paper liner helps to minimize stretching during installation.
Applications include dust and moisture seal for HVAC, appliances, gasketing, construction and electronic and transportation equipment.
Frequently Bought Together