Double Sided Applicator Brushes, Anti-Static, Gordon Brush
These brushes have two sides with two end styles (tapered and flat) to fit everybody's needs.
- Great alternative to hand-trimming acid brushes
- Available with stainless steel or zinc plated handle
- One side has a short flat end, and the other end has an extrastiff chiseled trim
Fibers are mechanically clamped into the handle for maximum fiber retention so that no adhesives can leach out and contaminate surfaces being cleaned.
Frequently Bought Together