Student Laboratory Coats
Supplier: DR INSTRUMENTS INC. SE
About this item
Fine quality lab coats for use in the classroom or laboratory by students.
- Side hand access
- Soft and comfortable fabric
- Perfect for use by elementary or middle school students
- Unisex kids lab coats - 60% cotton and 40% polyester twill fabric
These coats are both convenient and comfortable for student use. Perfect for use in any laboratory and offering exceptional body protection from spills and leaks.
4 Options Available Below
Product Details & Documents
Fine quality lab coats for use in the classroom or laboratory by students.
- Side hand access
- Soft and comfortable fabric
- Perfect for use by elementary or middle school students
- Unisex kids lab coats - 60% cotton and 40% polyester twill fabric
These coats are both convenient and comfortable for student use. Perfect for use in any laboratory and offering exceptional body protection from spills and leaks.
Specifications for Product Family
Specifications
- Catalog No:
- 470310-436
- 470310-438
- 470310-440
- 470310-442
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 60% cotton and 40% polyester twill fabric
- 60% cotton and 40% polyester twill fabric
- 60% cotton and 40% polyester twill fabric
- 60% cotton and 40% polyester twill fabric
Specifications
Product Family Options
Product Information
- Clothing SizeAvailabilityPrice
- Supplier Number: DR101-7Specifications:
More
Specifications
Cat. No.470310-436Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric60% cotton and 40% polyester twill fabric - Supplier Number: DR101-810Specifications:
More
Specifications
Cat. No.470310-438Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric60% cotton and 40% polyester twill fabric - Supplier Number: DR101-1214Specifications:
More
Specifications
Cat. No.470310-440Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric60% cotton and 40% polyester twill fabric - Supplier Number: DR101-1618Specifications:
More
Specifications
Cat. No.470310-442Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric60% cotton and 40% polyester twill fabric