Spatulas, Miniature, Plastic Handle, Excelta Corp®
Catálogo # 76250-658
Proveedor: EXCELTA CORPORATION
Especificaciones
- Color:Black handle
- Descripción:Miniature spatula, straight, 3 star quality, plastic handle, radius tip
- Longitud:62.5 mm (2.5")
- Matriz:Carbon steel, nickel plated
- Referencia:76250-658
- Blade Width:0.20 mm (0.008")
Especificaciones
Acerca de este artículo
These miniature spatulas are designed for placing epoxy/lubricants in very small amounts in a precise location.
- Miniature design for accessibility under scope
- Color coded width sizes for identification
- Different tip widths to fit different applications
These mini-spatula (oiler) are 2.5" long with a plastic handle. Mini-spatulas are ideal for placing epoxies or lubricant in very small amounts into tight places. They feature different size tips (color coated for identification).
Spatula shape: Flat end blade/straight handle.