Orders LinkContactUsLinkComponent
Dispensing Tips, Tapered, Jensen Tools
Dispensing Tips, Tapered, Jensen Tools
25, NT Premium Tapered, Red
Catalog # 76246-802
Supplier:   JENSEN GLOBAL INC
Número de proveedor #:  JG25-1.25HPTTX
Dispensing Tips, Tapered, Jensen Tools
Catalog # 76246-802
Supplier:  JENSEN GLOBAL INC
Dispensing Tips, Tapered, Jensen Tools
25, NT Premium Tapered, Red
Price
$1,399.98
Log in Para disponibilidad
Each (50UN) Paquete de 50
Product Details & Documents
Dispensing tip manufactured using the highest quality standards and the tightest possible tolerances.
  • UV-blocking additive
  • Scratch-free protection for working with sensitive parts
NT Premium tapered tips have an all-plastic tapered design to reduce backpressure and work best with medium to high viscosity materials.
Especificaciones
  • Color:
    Red
  • Point Style:
    NT Premium Tapered
  • Gauge:
    25